2,2'-Dithiodi(benzoic acid) ≥96.0%
Supplier: TCI America
Synonyms:
2,2'-Dithiosalicylic acid, 2,2'-Dithiodibenzoic acid
D1437-25G
D1437-500G
D1437-100G
TCD1437-025GEA
155.41
CAD
TCD1437-025G
TCD1437-500G
TCD1437-100G
2,2'-Dithiodi(benzoic acid) ≥96.0%
2,2'-Dithiodi(benzoic acid)
CAS Number: 119-80-2
MDL Number: MFCD00002465
Molecular Formula: C14H10O4S2
Molecular Weight: 306.35
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Sulfenylation
Other Product Names / Synonyms: Dithiosalicylic Acid
Formula:
C₁₄H₁₀O₄S₂ MW: 306.36 g/mol |
MDL Number:
MFCD00002465 CAS Number: 119-80-2 EINECS: 204-352-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...