(+)-Di-p-toluoyl-D-tartaric acid 98,0

Supplier: TCI America

Synonyms: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

D1417-250G D1417-25G
TCD1417-250GEA 224.36 CAD
TCD1417-250G TCD1417-025G
(+)-Di-p-toluoyl-D-tartaric acid 98,0
(+)-Di-p-toluoyl-D-tartaric acid

CAS Number: 32634-68-7
MDL Number: MFCD00008552
Molecular Formula: C20H18O8
Molecular Weight: 386.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 171
Specific rotation [a]20/D: 139 deg (C=1, EtOH)

Formula: C₂₀H₁₈O₈
MW: 386.36 g/mol
Melting Pt: 171 °C
MDL Number: MFCD00008552
CAS Number: 32634-68-7
EINECS: 251-132-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c46f0b55858bea3fdc79b3c28d9f6aadae05cde5&vid=c46f0b55858bea3fdc79b3c28d9f6aadae05cde5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR