Sodium dodecyl sulfate (SDS) ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D1403-25G D1403-500G
TCD1403-025GEA 25.72 CAD
TCD1403-025G TCD1403-500G
Sodium dodecyl sulfate (SDS) ≥97.0% (by GC, titration analysis)
Sodium dodecyl sulphate (SDS)

CAS Number: 151-21-3
MDL Number: MFCD00036175
Molecular Formula: C12H26O4S
Molecular Weight: 288.38
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 207
Flash Point (°C): 10


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: Dodecylsulfuric Acid Sodium Salt, Laurylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate, SDS

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f0929c97ab1c86d41ac28c906e7b67b678873f&vid=071d621c42ae42a23801f880f3a9fed48ab461e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR