3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥98.0%
Supplier: TCI America
Synonyms:
Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol
D1399-5G
D1399-25G
TCD1399-005GEA
112.18
CAD
TCD1399-005G
TCD1399-025G
3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥98.0%
3,4-Dihydroxy-3-cyclobutene-1,2-dione
CAS Number: 2892-51-5
MDL Number: MFCD00001334
Molecular Formula: C4H2O4
Molecular Weight: 114.06
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 190
Other Product Names / Synonyms: 1-Cyclobutene-3,4-dione-1,2-diol, Squaric Acid
Formula:
C₄H₂O₄ Melting Pt: 293 °C Density: 1 g/cm³ (20 °C) Flash Pt: 190 °C |
MDL Number:
MFCD00001334 CAS Number: 2892-51-5 EINECS: 220-761-4 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...