(+)-Dibenzoyl-D-tartaric acid monohydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D1398-500G
D1398-25G
TCD1398-500GEA
624.73
CAD
TCD1398-500G
TCD1398-025G
(+)-Dibenzoyl-D-tartaric acid monohydrate ≥98.0% (by titrimetric analysis)
(+)-Dibenzoyl-D-tartaric acid monohydrate
CAS Number: 80822-15-7
MDL Number: MFCD00150722
Molecular Formula: C18H14O8
Molecular Weight: 358.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 156
Specific rotation [a]20/D: 109 deg (C=1, EtOH)
Formula:
C₁₈H₁₄O₈·1H₂O Melting Pt: 156 °C |
MDL Number:
MFCD00150722 CAS Number: 80822-15-7 EINECS: 241-097-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...