1,1-Diphenylacetone ≥99.0%

Supplier: TCI America

Synonyms: asym. Diphenylacetone, 1,1-Diphenyl-2-propanone

D1397-10G D1397-25G
TCD1397-010GEA 82.16 CAD
TCD1397-010G TCD1397-025G
1,1-Diphenylacetone ≥99.0%
1,1-Diphenylacetone

CAS Number: 781-35-1
MDL Number: MFCD00008759
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 62


Other Product Names / Synonyms: 1,1-Diphenyl-2-propanone

Formula: (C₆H₅)₂CHC(O)CH₃
MW: 210.28 g/mol
Boiling Pt: 135 °C
Melting Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008759
CAS Number: 781-35-1
EINECS: 212-307-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50aecbc608591bddaab6e278cb78a095b1c0dda&vid=8345ebfa056d000383fa43d864f24be7552b646d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR