(-)-Di-p-toluoyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid

D1387-250G D1387-25G
TCD1387-250GEA 115.34 CAD
TCD1387-250G TCD1387-025G
(-)-Di-p-toluoyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)
(-)-Di-p-toluoyl-L-tartaric acid

CAS Number: 32634-66-5
MDL Number: MFCD00064440
Molecular Formula: C20H18O8
Molecular Weight: 386.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 172
Specific rotation [a]20/D: -139 deg (C=1, EtOH)

Formula: C₂₀H₁₈O₈
MW: 386.36 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00064440
CAS Number: 32634-66-5
EINECS: 251-131-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59b4b703a78699c7e653fd34eec7316407260c9c&vid=59b4b703a78699c7e653fd34eec7316407260c9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR