4-Chloro-2,6-dimethoxypyrimidine ≥98.0%

Supplier: TCI America

D1385-1G D1385-5G
TCD1385-1GEA 67.94 CAD
TCD1385-1G TCD1385-005G
4-Chloro-2,6-dimethoxypyrimidine ≥98.0%
4-Chloro-2,6-dimethoxypyrimidine

CAS Number: 6320-15-6
MDL Number: MFCD00006068
Molecular Formula: C6H7ClN2O2
Molecular Weight: 174.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 74

Formula: C₆H₇ClN₂O₂
Melting Pt: 74 °C
MDL Number: MFCD00006068
CAS Number: 6320-15-6
EINECS: 228-669-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR