(±)-2-Decanol ≥98.0%

Supplier: TCI America

D1379-5ML D1379-25ML
TCD1379-005MLEA 145.36 CAD
TCD1379-005ML TCD1379-025ML
(±)-2-Decanol ≥98.0%
(±)-2-Decanol

CAS Number: 1120-06-5
MDL Number: MFCD00004594
Molecular Formula: C10H22O
Molecular Weight: 158.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Melting point (°C): -4
Flash Point (°C): 80
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: 2-Decyl Alcohol, Methyl-n-octylcarbinol

Formula: C₁₀H₂₂O
Boiling Pt: 211 °C
Melting Pt: –4 °C
Density: 0.83 (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00004594
CAS Number: 1120-06-5
EINECS: 214-296-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24fc5195d7f1b8a9f21d7d40fe97096f1d6d1914&vid=3f2d4211427d52537718c968cb5d33940a66de6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR