Dioctyltin oxide ≥98.0% (by gravimetric analysis)
Supplier: TCI America
D1373-25G
D1373-500G
TCD1373-025GEA
64.78
CAD
TCD1373-025G
TCD1373-500G
Dioctyltin oxide ≥98.0% (by gravimetric analysis)
Dioctyltin oxide
CAS Number: 870-08-6
MDL Number: MFCD00013839
Molecular Formula: C16H34OSn
Molecular Weight: 361.16
Purity/Analysis Method: >98.0% (W)
Form: Crystal
Formula:
C₁₆H₃₄OSn Boiling Pt: 230 °C (1013 hPa) Density: 1.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013839 CAS Number: 870-08-6 EINECS: 212-791-1 UN: 3146 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...