N,N-Diethylacetoacetamide ≥98.0%

Supplier: TCI America

D1371-500G D1371-25G
TCD1371-500GEA 554.58 CAD
TCD1371-500G TCD1371-025G
N,N-Diethylacetoacetamide ≥98.0%
N,N-Diethylacetoacetamide

CAS Number: 2235-46-3
MDL Number: MFCD00026728
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 120
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: N-Acetoacetyldiethylamine

Formula: C₈H₁₅NO₂
Boiling Pt: 116 °C
Density: 1.00 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00026728
CAS Number: 2235-46-3
EINECS: 218-792-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4001bbc98c9beb322bafa71efd3d406c7c90e99f&vid=4001bbc98c9beb322bafa71efd3d406c7c90e99f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR