Dipentaerythritol ≥80.0%

Supplier: TCI America

Synonyms: 2,2'-(Oxybis(methylene))bis(2-(hydroxymethyl)propane-1,3-diol)

D1342-25G D1342-500G
TCD1342-025GEA 30.02 CAD
TCD1342-025G TCD1342-500G
Dipentaerythritol ≥80.0%
Dipentaerythritol

CAS Number: 126-58-9
MDL Number: MFCD00004691
Molecular Formula: C10H22O7
Molecular Weight: 254.28
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Melting point (°C): 218
Flash Point (°C): 200


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bis[2,2,2-tris(hydroxymethyl)ethyl] Ether

Formula: C₁₀H₂₂O₇
MW: 254.28 g/mol
Melting Pt: 218 °C
Flash Pt: 200 °C
MDL Number: MFCD00004691
CAS Number: 126-58-9
EINECS: 204-794-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4a53ab35a1046d0c1212ca77e079439882d75bf&vid=a4a53ab35a1046d0c1212ca77e079439882d75bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR