2,2-Dimethylglutaric anhydride 98,0

Supplier: TCI America

Synonyms: 3,3-Dimethyldihydro-2H-pyran-2,6(3H)-dione

D1325-5G D1325-25G
TCD1325-5GEA 246.48 CAD
TCD1325-5G TCD1325-025G
2,2-Dimethylglutaric anhydride 98,0
2,2-Dimethylglutaric anhydride

CAS Number: 2938-48-9
MDL Number: MFCD00006682
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 38
Flash Point (°C): 113

Formula: C₇H₁₀O₃
MW: 142.1525 g/mol
Boiling Pt: 270 °C
Melting Pt: 38 °C
Flash Pt: 113 °C
MDL Number: MFCD00006682
CAS Number: 2938-48-9
EINECS: 220-922-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7828fe974864f68a7368808ed5ad070c6a53e47&vid=c7828fe974864f68a7368808ed5ad070c6a53e47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR