3,3-Dimethylglutaric anhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4,4-Dimethyldihydro-2H-pyran-2,6(3H)-dione

D1323-5G D1323-25G
TCD1323-005GEA 121.66 CAD
TCD1323-005G TCD1323-025G
3,3-Dimethylglutaric anhydride ≥97.0% (by titrimetric analysis)
3,3-Dimethylglutaric anhydride

CAS Number: 4160-82-1
MDL Number: MFCD00006684
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 126

Formula: C₇H₁₀O₃
MW: 142.15 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00006684
CAS Number: 4160-82-1
EINECS: 223-998-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d379f5c53bb0432499187c3031ff388dc9cf3e20&vid=d379f5c53bb0432499187c3031ff388dc9cf3e20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR