1,1'-Dimethylferrocene ≥93.0%

Supplier: TCI America

D1273-25G D1273-5G
TCD1273-25GEA 741.02 CAD
TCD1273-25G TCD1273-005G
1,1'-Dimethylferrocene ≥93.0%
1,1'-Dimethylferrocene

CAS Number: 1291-47-0
MDL Number: MFCD00001425
Molecular Formula: C12H14Fe
Molecular Weight: 214.09
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 40
Flash Point (°C): 108

Formula: C₁₂H₁₄Fe
Melting Pt: 40 °C
Flash Pt: 108 °C
MDL Number: MFCD00001425
CAS Number: 1291-47-0
EINECS: 215-067-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a3103e0e5b4d2721c12eeb92e5d4ba29aeb39e6&vid=7a3103e0e5b4d2721c12eeb92e5d4ba29aeb39e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR