2,4-Dimethylquinoline ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D1237-1G D1237-5G
TCD1237-001GEA 226.41 CAD
TCD1237-001G TCD1237-005G
2,4-Dimethylquinoline ≥98.0% (by titrimetric analysis)
2,4-Dimethylquinoline

CAS Number: 1198-37-4
MDL Number: MFCD00006760
Molecular Formula: C11H11N
Molecular Weight: 157.22
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 159
Flash Point (°C): 110
Specific Gravity (20/20): 1.05

Formula: C₁₁H₁₁N
Boiling Pt: 159 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00006760
CAS Number: 1198-37-4
EINECS: 214-832-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2641cb9e098a22f72d5bbf49e5d97a6e75b686d&vid=b1941df64b3d1238f85cb6df86c4afe5de40882e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR