4,4'-Dimethylbiphenyl ≥97.0%

Supplier: TCI America

Synonyms: 4,4'-Bitolyl, 4,4'-Dimethyldiphenyl

D1234-1G D1234-5G D1234-25G
TCD1234-001GEA 33.18 CAD
TCD1234-001G TCD1234-005G TCD1234-25G
4,4'-Dimethylbiphenyl ≥97.0%
4,4'-Dimethylbiphenyl

CAS Number: 613-33-2
MDL Number: MFCD00008545
Molecular Formula: C14H14
Molecular Weight: 182.27
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 122


Other Product Names / Synonyms: 4,4'-Bitolyl

Formula: C₁₄H₁₄
MW: 182.27 g/mol
Boiling Pt: 295
Melting Pt: 122 °C
Density: 1.102 g/cm³ (20 °C)
MDL Number: MFCD00008545
CAS Number: 613-33-2
EINECS: 210-337-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d10d755679b49455b5b2ed5bec69515ab03531cb&vid=d10d755679b49455b5b2ed5bec69515ab03531cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR