Dihydrocoumarin ≥98.0%

Supplier: TCI America

Synonyms: Chroman-2-One, 1,2-Benzodihydropyrone

D1223-25G D1223-500G
TCD1223-025GEA 33.18 CAD
TCD1223-025G TCD1223-500G
Dihydrocoumarin ≥98.0%
Dihydrocoumarin

CAS Number: 119-84-6
MDL Number: MFCD00006881
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 149
Flash Point (°C): 130
Freezing point (°C): 22

Formula: C₉H₈O₂
Boiling Pt: 149 °C
Flash Pt: 130 °C
MDL Number: MFCD00006881
CAS Number: 119-84-6
EINECS: 204-354-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aff8a3685fd581f83497aae634a03a019a52181c&vid=aff8a3685fd581f83497aae634a03a019a52181c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR