1-Dodecansulfonic acid sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1222-5G D1222-25G
TCD1222-005GEA 94.17 CAD
TCD1222-005G TCD1222-025G
1-Dodecansulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
1-Dodecansulphonic acid sodium salt

CAS Number: 2386-53-0
MDL Number: MFCD00007527
Molecular Formula: C12H26O3S
Molecular Weight: 272.38
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: 1-Dodecanesulfonic Acid Sodium Salt, Laurylsulfonic Acid Sodium Salt

Formula: C₁₂H₂₅NaO₃S
Melting Pt: 300 °C
Density: 1.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0
EINECS: 219-200-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR