2,5-Dibromopyridine ≥99.0%
Supplier: TCI America
|
Warning
|
D1217-500G
D1217-5G
D1217-25G
TCD1217-500GEA
1017.52
CAD
TCD1217-500G
TCD1217-005G
TCD1217-025G
2,5-Dibromopyridine ≥99.0%
2,5-Dibromopyridine
CAS Number: 624-28-2
MDL Number: MFCD00006221
Molecular Formula: C5H3Br2N
Molecular Weight: 236.89
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 93
Formula:
C₅H₃Br₂N MW: 236.89 g/mol Boiling Pt: 235 °C Melting Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006221 CAS Number: 624-28-2 EINECS: 210-839-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...