Sodium 1-decanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1181-25G D1181-5G
TCD1181-025GEA 133.38 CAD
TCD1181-025G TCD1181-5G
Sodium 1-decanesulfonate ≥98.0% (by titrimetric analysis)
Sodium 1-decanesulphonate

CAS Number: 13419-61-9
MDL Number: MFCD00007526
Molecular Formula: C10H22O3S
Molecular Weight: 244.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1-Decanesulfonic Acid Sodium Salt

Formula: C₁₀H₂₁NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6225ac831d49953c0f9e1877c73cefe0c3616bd&vid=e6225ac831d49953c0f9e1877c73cefe0c3616bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR