Dibutyl sulfoxide ≥96.0%

Supplier: TCI America

D1173-25G D1173-5G
TCD1173-025GEA 246.32 CAD
TCD1173-025G TCD1173-5G
Dibutyl sulfoxide ≥96.0%
Dibutyl sulphoxide

CAS Number: 2168-93-6
MDL Number: MFCD00002093
Molecular Formula: C8H18OS
Molecular Weight: 162.29
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Flash Point (°C): 121
Freezing point (°C): 34


Other Product Names / Synonyms: Butyl Sulfoxide

Formula: C₈H₁₈OS
Flash Pt: 121 °C
MDL Number: MFCD00002093
CAS Number: 2168-93-6
EINECS: 218-511-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac8d6bd3df4619cdaa1c0689eb1d30b95cad4c2c&vid=ac8d6bd3df4619cdaa1c0689eb1d30b95cad4c2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR