3,5-Dichlorosalicylaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde

D1170-25G D1170-5G
TCD1170-025GEA 122.18 CAD
TCD1170-025G TCD1170-5G
3,5-Dichlorosalicylaldehyde ≥98.0%
3,5-Dichlorosalicylaldehyde

CAS Number: 90-60-8
MDL Number: MFCD00003320
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 94


Other Product Names / Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde

Formula: C₇H₄Cl₂O₂
Boiling Pt: 723 °C (1013 hPa)
Melting Pt: 94 °C
MDL Number: MFCD00003320
CAS Number: 90-60-8
EINECS: 202-005-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d954b3cbbab20c71a45d9f65b00e5c5e582eafe&vid=6d954b3cbbab20c71a45d9f65b00e5c5e582eafe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR