2,5-Dimethyl-2,5-hexanediol ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,1,4,4-Tetramethyl-1,4-butanediol
D1167-25G
D1167-500G
TCD1167-025GEA
36.34
CAD
TCD1167-025G
TCD1167-500G
2,5-Dimethyl-2,5-hexanediol ≥99.0%
2,5-Dimethyl-2,5-hexanediol
CAS Number: 110-03-2
MDL Number: MFCD00004473
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 89
Flash Point (°C): 127
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,5-Dihydroxy-2,5-dimethylhexane
Formula:
C₈H₁₈O₂ Boiling Pt: 127 °C Melting Pt: 89 °C Density: 0.898 g/cm³ (20 °C) Flash Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004473 CAS Number: 110-03-2 EINECS: 203-731-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...