3,5-Dimethoxybenzaldehyde ≥98.0%
Supplier: TCI America
D1164-25G
D1164-5G
TCD1164-025GEA
325.48
CAD
TCD1164-025G
TCD1164-005G
3,5-Dimethoxybenzaldehyde ≥98.0%
3,5-Dimethoxybenzaldehyde
CAS Number: 7311-34-4
MDL Number: MFCD00003366
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 151
Melting point (°C): 47
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 151 °C Melting Pt: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003366 CAS Number: 7311-34-4 EINECS: 230-772-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...