3,4-Diaminobenzoic acid

Supplier: TCI America
Warning

D1117-25G D1117-5G
TCD1117-25GEA 88.48 CAD
TCD1117-25G TCD1117-005G
3,4-Diaminobenzoic acid
3,4-Diaminobenzoic acid

CAS Number: 619-05-6
MDL Number: MFCD00007726
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Form: Crystal
Color: Yellow
Melting point (°C): 208

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 208 °C
Density: 1.544 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6
EINECS: 210-577-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5228867adcc1c2a4d393f4801214e157ab6f565e&vid=324b0cf5cb0bd2f43d60affd468c5979ba354c34&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR