Dodecylbenzene ≥98.0%
Supplier: TCI America
Synonyms:
n-Dodecylbenzene, 1-Phenyldodecane
D1074-25ML
D1074-500ML
TCD1074-025MLEA
156.91
CAD
TCD1074-025ML
TCD1074-500ML
Dodecylbenzene ≥98.0%
Dodecylbenzene
CAS Number: 123-01-3
MDL Number: MFCD00008974
Molecular Formula: C18H30
Molecular Weight: 246.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Melting point (°C): 3
Flash Point (°C): 124
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Laurylbenzene, 1-Phenyldodecane
Formula:
C₁₈H₃₀ Boiling Pt: 148 °C Melting Pt: 3 °C Density: 0.86 (20 °C) Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008974 CAS Number: 123-01-3 EINECS: 204-591-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...