2,3-Diaminonaphthalene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D1045-1G
D1045-5G
TCD1045-001GEA
213.84
CAD
TCD1045-001G
TCD1045-005G
2,3-Diaminonaphthalene ≥98.0% (by titrimetric analysis)
2,3-Diaminonaphthalene
CAS Number: 771-97-1
MDL Number: MFCD00004116
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 198
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 2,3-Naphthalenediamine
Formula:
C₁₀H₁₀N₂ Boiling Pt: 343 °C (1013 hPa) Melting Pt: 198 °C Density: 1.0968 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004116 CAS Number: 771-97-1 EINECS: 212-241-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...