1,4-Dicyclohexylbenzene ≥98.0%

Supplier: TCI America

D1028-250G D1028-25G
TCD1028-250GEA 881.64 CAD
TCD1028-250G TCD1028-025G
1,4-Dicyclohexylbenzene ≥98.0%
1,4-Dicyclohexylbenzene

CAS Number: 1087-02-1
MDL Number: MFCD00001452
Molecular Formula: C18H26
Molecular Weight: 242.41
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 102

Formula: C₁₈H₂₆
Boiling Pt: 195 °C
Melting Pt: 102 °C
Density: 1.049 g/cm³ (0 °C)
MDL Number: MFCD00001452
CAS Number: 1087-02-1
EINECS: 214-121-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e914a910a0684b2d33953e2095107ecb6698cc8c&vid=f24f9776aedaaa3d8168c89bffeb607be571dac4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR