Diphenyl sulfoxide ≥99.0%
Supplier: TCI America
D1002-5G
D1002-25G
D1002-100G
TCD1002-5GEA
33.18
CAD
TCD1002-5G
TCD1002-025G
TCD1002-100G
Diphenyl sulfoxide ≥99.0%
Diphenyl sulphoxide
CAS Number: 945-51-7
MDL Number: MFCD00002085
Molecular Formula: C12H10OS
Molecular Weight: 202.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 198
Melting point (°C): 71
Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Phenyl Sulfoxide
Formula:
C₁₂H₁₀OS Boiling Pt: 198 °C Melting Pt: 71 °C |
MDL Number:
MFCD00002085 CAS Number: 945-51-7 EINECS: 213-415-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...