n-Dotriacontane ≥96.0%
Supplier: TCI America
D0998-25G
D0998-5G
TCD0998-25GEA
356.91
CAD
TCD0998-25G
TCD0998-005G
n-Dotriacontane ≥96.0%
n-Dotriacontane
CAS Number: 544-85-4
MDL Number: MFCD00009411
Molecular Formula: C32H66
Molecular Weight: 450.88
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 70
Flash Point (°C): 243
Formula:
H₃C(CH₂)₃₀CH₃ Boiling Pt: 220 °C Melting Pt: 70 °C Density: 0.7798 g/cm³ (20 °C) Flash Pt: 243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009411 CAS Number: 544-85-4 EINECS: 208-881-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...