Laurinaldehyde ≥95.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

D0979-25ML D0979-500ML
TCD0979-025MLEA 99.68 CAD
TCD0979-025ML TCD0979-500ML
Laurinaldehyde ≥95.0% stabilized
Laurinaldehyde

(stabilized with DL-alpha-Tocopherol)
CAS Number: 112-54-9
MDL Number: MFCD00007017
Molecular Formula: C12H24O
Molecular Weight: 184.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 241
Flash Point (°C): 82
Freezing point (°C): 12
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Dodecanaldehyde, Dodecyl Aldehyde, Laurinaldehyde, Lauryl Aldehyde

Formula: C₁₂H₂₄O
Boiling Pt: 241 °C
Melting Pt: 44 °C
Density: 0.83 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007017
CAS Number: 112-54-9
EINECS: 203-983-6
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e9d778efc20cc8119a0ab4c4092abddceabef93&vid=6e9d778efc20cc8119a0ab4c4092abddceabef93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR