1,12-Dodecanediol ≥99.0%

Supplier: TCI America

D0969-25G D0969-500G D0969-100G
TCD0969-025GEA 59.95 CAD
TCD0969-025G TCD0969-500G TCD0969-100G
1,12-Dodecanediol ≥99.0%
1,12-Dodecanediol

CAS Number: 5675-51-4
MDL Number: MFCD00004755
Molecular Formula: C12H26O2
Molecular Weight: 202.34
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 189
Melting point (°C): 82
Flash Point (°C): 176


Other Product Names / Synonyms: 1,12-Dihydroxydodecane, Dodecamethylene Glycol

Formula: C₁₂H₂₆O₂
Boiling Pt: 189 °C
Melting Pt: 82 °C
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 176 °C
MDL Number: MFCD00004755
CAS Number: 5675-51-4
EINECS: 227-133-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f90903abbae6e295b346465f214a3e42083424f&vid=0f90903abbae6e295b346465f214a3e42083424f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR