Behenic acid ≥80.0%

Supplier: TCI America

D0963-25G D0963-500G
TCD0963-025GEA 37.92 CAD
TCD0963-025G TCD0963-500G
Behenic acid ≥80.0%
Behenic acid

CAS Number: 112-85-6
MDL Number: MFCD00002807
Molecular Formula: C22H44O2
Molecular Weight: 340.59
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Boiling point (°C): 306
Melting point (°C): 79
Flash Point (°C): 228


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Docosanoic Acid

Formula: C₂₂H₄₄O₂
Boiling Pt: 306 °C
Melting Pt: 79 °C
Density: 0.8221 g/cm³ (20 °C)
Flash Pt: 228 °C
Storage Temperature: Ambient
MDL Number: MFCD00002807
CAS Number: 112-85-6
EINECS: 204-010-8
Merck Index: 13,01024
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de551e8de9d7b0d41fea2a81ebbb7808da03f2eb&vid=b9c03d2c163fc53db6c2f90a1ec4f4d80ab7c004&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR