n-Docosane ≥98.0%

Supplier: TCI America

D0962-25G D0962-500G D0962-100G
TCD0962-025GEA 88.43 CAD
TCD0962-025G TCD0962-500G TCD0962-100G
n-Docosane ≥98.0%
n-Docosane

CAS Number: 629-97-0
MDL Number: MFCD00009348
Molecular Formula: C22H46
Molecular Weight: 310.61
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 380
Melting point (°C): 46
Flash Point (°C): 166

Formula: H₃C(CH₂)₂₀CH₃
Boiling Pt: 380 °C
Melting Pt: 46 °C
Density: 0.7944 g/cm³ (20 °C)
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00009348
CAS Number: 629-97-0
EINECS: 211-121-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3635f314f71394a8f43529e76eb32b079404d88e&vid=f2f3d901bd6047ce16d7836325fcf93fe2bd15b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR