2,6-Dihydroxynaphthalene ≥95.0%

Supplier: TCI America
Warning

D0956-5G D0956-25G
TCD0956-005GEA 83.74 CAD
TCD0956-005G TCD0956-25G
2,6-Dihydroxynaphthalene ≥95.0%
2,6-Dihydroxynaphthalene

CAS Number: 581-43-1
MDL Number: MFCD00004082
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 222


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,6-Naphthalenediol

Formula: C₁₀H₈O₂
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 222 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004082
CAS Number: 581-43-1
EINECS: 209-465-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6949915e813308448f2b8b2397d06d507770656e&vid=6949915e813308448f2b8b2397d06d507770656e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR