3,3'-Dithiodipropionic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,3-Dithiodipropionic Acid
D0947-25G
D0947-500G
D0947-100G
TCD0947-025GEA
36.94
CAD
TCD0947-025G
TCD0947-500G
TCD0947-100G
3,3'-Dithiodipropionic acid ≥99.0% (by GC, titration analysis)
3,3'-Dithiodipropionic acid
CAS Number: 1119-62-6
MDL Number: MFCD00002780
Molecular Formula: C6H10O4S2
Molecular Weight: 210.26
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 157
Formula:
C₆H₁₀O₄S₂ MW: 210.28 g/mol Melting Pt: 157 °C |
MDL Number:
MFCD00002780 CAS Number: 1119-62-6 EINECS: 214-284-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...