5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 98,0

Supplier: TCI America
Warning

Synonyms: DTNB, Ellman's reagent, 5,5'-Disulfanediylbis(2-nitrobenzoic acid), 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid

D0944-5G D0944-25G D0944-1G
TCD0944-5GEA 150.16 CAD
TCD0944-5G TCD0944-025G TCD0944-001G
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 98,0
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

[for Determination of SH groups]
CAS Number: 69-78-3
MDL Number: MFCD00007140
Molecular Formula: C14H8N2O8S2
Molecular Weight: 396.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Functionality Creating Reagents
Other Product Names / Synonyms: Bis(3-carboxy-4-nitrophenyl) Disulfide, DTNB, Ellman's Reagent

Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00007140
CAS Number: 69-78-3
EINECS: 200-714-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfbe9808c3b09107460c79491cc6aed807397053&vid=d38ba17ab0def6bd27da9d0a8d722876da8eb746&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR