2,6-Di-tert-butylphenol ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Di(dimethylethyl)-2-hydroxybenzene

D0941-25G D0941-500G
TCD0941-025GEA 40.02 CAD
TCD0941-025G TCD0941-500G
2,6-Di-tert-butylphenol ≥98.0%
2,6-Di-tert-butylphenol

CAS Number: 128-39-2
MDL Number: MFCD00008820
Molecular Formula: C14H22O
Molecular Weight: 206.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 253
Flash Point (°C): 118
Freezing point (°C): 37

Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 253 °C
Melting Pt: 34…37 °C
Density: 0.914 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00008820
CAS Number: 128-39-2
EINECS: 204-884-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2d34f096ecb707e3dfc9ec630028569bcaa8829&vid=ac0c2ae9fb7e5be152c2b638f99e96a08b4a2a81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR