2,5-Di-tert-butylhydroquinone ≥98.0%

Supplier: TCI America

Synonyms: BHQ

D0940-25G D0940-500G
TCD0940-025GEA 33.75 CAD
TCD0940-025G TCD0940-500G
2,5-Di-tert-butylhydroquinone ≥98.0%
2,5-Di-tert-butylhydroquinone

CAS Number: 88-58-4
MDL Number: MFCD00008825
Molecular Formula: C14H22O2
Molecular Weight: 222.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 321
Melting point (°C): 215
Flash Point (°C): 178

Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Boiling Pt: 321 °C
Melting Pt: 215 °C
Density: 1.078 g/cm³ (20 °C)
Flash Pt: 178 °C
Storage Temperature: Ambient
MDL Number: MFCD00008825
CAS Number: 88-58-4
EINECS: 201-841-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fad0297e4572a20354bb96a211949e410372c1&vid=a3fad0297e4572a20354bb96a211949e410372c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR