Di-isopropanolamine ≥90.0%

Supplier: TCI America
Warning

D0924-500G D0924-25G
TCD0924-500GEA 71.1 CAD
TCD0924-500G TCD0924-25G
Di-isopropanolamine ≥90.0%
Di-isopropanolamine

CAS Number: 110-97-4
MDL Number: MFCD00004531
Molecular Formula: C6H15NO2
Molecular Weight: 133.19
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Boiling point (°C): 249
Flash Point (°C): 127
Freezing point (°C): 33


Other Product Names / Synonyms: Iminodipropanol

Formula: C₆H₁₅NO₂
Boiling Pt: 249 °C
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00004531
CAS Number: 110-97-4
EINECS: 203-820-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86658a4b7d6435240992d93c829b858b747a216b&vid=86658a4b7d6435240992d93c829b858b747a216b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR