Dibenzoylmethane ≥98.0% (by GC)
Supplier: TCI America
D0910-25G
D0910-500G
D0910-100G
TCD0910-025GEA
33.18
CAD
TCD0910-025G
TCD0910-500G
TCD0910-100G
Dibenzoylmethane ≥98.0% (by GC)
Dibenzoylmethane
CAS Number: 120-46-7
MDL Number: MFCD00003085
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 221
Melting point (°C): 78
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Dibenzoylmethane
Formula:
(C₆H₅)C(O)CH₂C(O)(C₆H₅) Boiling Pt: 221 °C Melting Pt: 78 °C Density: 0.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003085 CAS Number: 120-46-7 EINECS: 204-398-9 Merck Index: 13,03033 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...