Bathophenanthroline ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D0905-5G D0905-1G
TCD0905-005GEA 455.04 CAD
TCD0905-005G TCD0905-001G
Bathophenanthroline ≥99.0% (by titrimetric analysis)
Bathophenanthroline

CAS Number: 1662-01-7
MDL Number: MFCD00004976
Molecular Formula: C24H16N2
Molecular Weight: 332.41
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 221


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Analytical Science, Phenanthrolines (Chelating Agents)
Other Product Names / Synonyms: 4,7-Diphenyl-1,10-phenanthroline

Formula: C₂₄H₁₆N₂
Boiling Pt: 508 °C (1013 hPa)
Melting Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00004976
CAS Number: 1662-01-7
EINECS: 216-767-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77dde1e3336d48b30fd99e5b71e11ae508384335&vid=77dde1e3336d48b30fd99e5b71e11ae508384335&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR