2,5-Diphenyloxazole ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D0902-25G D0902-500G D0902-100G
TCD0902-025GEA 229.73 CAD
TCD0902-025G TCD0902-500G TCD0902-100G
2,5-Diphenyloxazole ≥99.0% (by HPLC, total nitrogen)
2,5-Diphenyloxazole

[for scintillation spectrometry]
CAS Number: 92-71-7
MDL Number: MFCD00005306
Molecular Formula: C15H11NO
Molecular Weight: 221.26
Purity/Analysis Method: >99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 72


Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: DPO, PPO

Formula: C₁₅H₁₁NO
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 72 °C
Density: 1.094 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b96dcdbb61120b87c52a3c9b5e8bba19e87ac710&vid=af2ab6cb1845b917f9171f581102b6ac401076a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR