Benzhydryl alcohol ≥99.0%

Supplier: TCI America

Synonyms: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

D0898-500G D0898-25G
TCD0898-500GEA 139.04 CAD
TCD0898-500G TCD0898-025G
Benzhydryl alcohol ≥99.0%
Benzhydryl alcohol

CAS Number: 91-01-0
MDL Number: MFCD00004488
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: Diphenylcarbinol, Diphenylmethanol

Formula: C₁₃H₁₂O
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 67 °C
Density: 1.054 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004488
CAS Number: 91-01-0
EINECS: 202-033-8
Merck Index: 12,01121
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d538f1edeb64975b5552258e8aee5a7c3b864b4d&vid=d538f1edeb64975b5552258e8aee5a7c3b864b4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR