1,1-Diphenylethylene ≥98.0% stabilised
Supplier: TCI America
Synonyms:
asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
Stabilization:
Stabilised with Stabilised
D0886-25G
D0886-5G
D0886-250G
TCD0886-025GEA
147.1
CAD
TCD0886-025G
TCD0886-005G
TCD0886-250G
1,1-Diphenylethylene ≥98.0% stabilised
1,1-Diphenylethylene
(stabilized with HQ)
CAS Number: 530-48-3
MDL Number: MFCD00008583
Molecular Formula: C14H12
Molecular Weight: 180.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 277
Flash Point (°C): 110
Freezing point (°C): 9
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C
Formula:
(C₆H₅)₂CCH₂ Boiling Pt: 277 °C Density: 1.03 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008583 CAS Number: 530-48-3 EINECS: 208-482-6 Merck Index: 13,03355 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...