N,N'-Diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D0878-5G
D0878-25G
TCD0878-005GEA
165.28
CAD
TCD0878-005G
TCD0878-025G
N,N'-Diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Diphenylbenzidine
CAS Number: 531-91-9
MDL Number: MFCD00003016
Molecular Formula: C24H20N2
Molecular Weight: 336.44
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 242
Flash Point (°C): 110
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Dianilinobiphenyl
Formula:
C₂₄H₂₀N₂ Melting Pt: 242 °C Flash Pt: 110 °C |
MDL Number:
MFCD00003016 CAS Number: 531-91-9 EINECS: 208-521-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...