Sodium diphenylamine-4-sulfonate 97,0

Supplier: TCI America

Synonyms: Diphenylamine-4-sulfonic acid, sodium salt, Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium Diphenylaminesulfonate, Sodium (4-anilinobenzenesulfonate), Sodium (4-anilinobenzenesulphonate), Sodium N-phenylsulfanilate, Diphenylamine-4-sulphonic acid, sodium salt

D0877-1G D0877-25G
TCD0877-001GEA 53.72 CAD
TCD0877-001G TCD0877-025G
Sodium diphenylamine-4-sulfonate 97,0
Sodium diphenylamine-4-sulphonate

CAS Number: 6152-67-6
MDL Number: MFCD00036468
Molecular Formula: C12H11NO3S
Molecular Weight: 271.27
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal


Other Product Names / Synonyms: Diphenylamine-4-sulfonic Acid Sodium Salt, N-Phenylsulfanilic Acid Sodium Salt

Formula: C₁₂H₁₀NNaO₃S
MW: 271.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efdfbb44de35ba215e453ac736fcd471db14b696&vid=efdfbb44de35ba215e453ac736fcd471db14b696&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR