Diphenylacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

D0869-25G D0869-500G D0869-100G
TCD0869-025GEA 67.21 CAD
TCD0869-025G TCD0869-500G TCD0869-100G
Diphenylacetic acid ≥98.0% (by HPLC, titration analysis)
Diphenylacetic acid

CAS Number: 117-34-0
MDL Number: MFCD00004251
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 146


Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction)
Other Product Names / Synonyms: Diphenylmethane-alpha-carboxylic Acid

Formula: C₁₄H₁₂O₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 146 °C
Density: 1.258 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004251
CAS Number: 117-34-0
EINECS: 204-185-0
Merck Index: 13,03348
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd00d90aa169575d080d96a1423236c561d7b7b&vid=0fd00d90aa169575d080d96a1423236c561d7b7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR