Diethyl diethylmalonate ≥97.0%

Supplier: TCI America

D0867-25ML D0867-500ML
TCD0867-025MLEA 33.18 CAD
TCD0867-025ML TCD0867-500ML
Diethyl diethylmalonate ≥97.0%
Diethyl diethylmalonate

CAS Number: 77-25-8
MDL Number: MFCD00009132
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 94
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Diethylmalonic Acid Diethyl Ester

Formula: C₁₁H₂₀O₄
Boiling Pt: 230 °C
Density: 0.99 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00009132
CAS Number: 77-25-8
EINECS: 201-016-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098b350ac12a0f7f1e008fb03c0ed58cd14467cc&vid=098b350ac12a0f7f1e008fb03c0ed58cd14467cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR